man

Test

CAS RN 108-77-0

ClNClNClN

CAS RN 108-77-0

IUPAC Name: trichloro-1,3,5-triazine
Synonyms: cyanuric chloride cyanuric acid chloride trichlorocyanidine cyanuric trichloride tricyanogen chloride cyanuryl chloride cyanuric acid trichloride sym-trichlorotriazine s-triazine trichloride
Molecular Weight (g/mol): 184.4
Molecular Formula: C3Cl3N3
InChi Key: MGNCLNQXLYJVJD-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=NC(Cl)=N1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)