man

Test

CAS RN 108-93-0

OH

CAS RN 108-93-0

IUPAC Name: cyclohexanol
Synonyms: cyclohexanol naxol hydrophenol cicloesanolo hydralin cykloheksanol hexalin adronol adronal Anol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: HPXRVTGHNJAIIH-UHFFFAOYSA-N
SMILES: OC1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)