man

Test

CAS RN 1080028-76-7

OHNO

CAS RN 1080028-76-7

IUPAC Name: N-(oxan-4-yl)oxan-4-amine
Synonyms: N-(oxan-4-yl)oxan-4-amine
Molecular Weight (g/mol): 185.27
Molecular Formula: C10H19NO2
InChi Key: WWPWWQFNGOWXOS-UHFFFAOYSA-N
SMILES: C1CC(CCO1)NC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results