man

Test

CAS RN 108307-56-8

FFFON

CAS RN 108307-56-8

IUPAC Name: 2-[3-(trifluoromethoxy)phenyl]acetonitrile
Synonyms: 2-[3-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: WZLPHJZVNJXHPV-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results