man

Test

CAS RN 1083326-28-6

BOOH3CCH3H3CH3CNNHSOO

CAS RN 1083326-28-6

IUPAC Name: N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
Synonyms: N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
Molecular Weight (g/mol): 360.24
Molecular Formula: C17H21BN2O4S
InChi Key: UXJVHVXONVGHIL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(NS(=O)(=O)C2=CC=CC=C2)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results