man

Test

CAS RN 108381-28-8

ONNCl

CAS RN 108381-28-8

IUPAC Name: 4-(benzyloxy)-2-chloropyrimidine
Synonyms: 4-(benzyloxy)-2-chloropyrimidine
Molecular Weight (g/mol): 220.66
Molecular Formula: C11H9ClN2O
InChi Key: SNCGVIVLUWJDQN-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(OCC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results