man

Test

CAS RN 108448-77-7

CH3H3CSOONH

CAS RN 108448-77-7

IUPAC Name: 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Synonyms: 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Molecular Weight (g/mol): 215.31
Molecular Formula: C10H17NO2S
InChi Key: DPJYJNYYDJOJNO-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC11CS(=O)(=O)NC1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results