CAS RN 108468-00-4
CAS RN 108468-00-4
IUPAC Name:
tert-butyl N-{[4-(aminomethyl)phenyl]methyl}carbamate
Synonyms:
tert-butyl N-{[4-(aminomethyl)phenyl]methyl}carbamate
Molecular Weight (g/mol):
236.32
Molecular Formula:
C13H20N2O2
InChi Key:
NUANLVJLUYWSER-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC1=CC=C(CN)C=C1
1
–
1
of
1
results
1
1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene, 90%, Thermo Scientific Chemicals
CAS: 108468-00-4 Molecular Formula: C13H20N2O2 Molecular Weight (g/mol): 236.31 MDL Number: MFCD02683058 InChI Key: NUANLVJLUYWSER-UHFFFAOYSA-N Synonym: 1-n-boc-aminomethyl-4-aminomethyl benzene,tert-butyl 4-aminomethyl benzylcarbamate,tert-butyl n-4-aminomethyl benzyl carbamate,4-aminomethyl-benzyl-carbamic acid tert-butyl ester,tert-butyl n-4-aminomethyl phenyl methyl carbamate,n-4-aminomethyl phenyl methyl tert-butoxy carboxamide,n-boc-p-xylylenediamine,4-tert-butoxycarbonylaminomethyl benzylamine PubChem CID: 3354775 IUPAC Name: tert-butyl N-[[4-(aminomethyl)phenyl]methyl]carbamate SMILES: CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CN