accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 31202-69-4

CAS RN 31202-69-4

IUPAC Name: 2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: 2-amino-6-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 246.31
Molecular Formula: C11H22N2O4
InChi Key: VVQIIIAZJXTLRE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(N)C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.