accessibility menu, dialog, popup

UserName

CAS RN 66168-50-1

NH2NNOOH(?)

CAS RN 66168-50-1

IUPAC Name: 4-[amino(nitroso)methylidene]-1,4-dihydropyridin-1-ol
Synonyms: 4-[amino(nitroso)methylidene]pyridin-1-ol
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: DYPGYCQXSLMOIR-UHFFFAOYSA-N
SMILES: NC(N=O)=C1C=CN(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results