accessibility menu, dialog, popup

UserName

CAS RN 66315-23-9

H3COOHNCH3NH2

CAS RN 66315-23-9

IUPAC Name: ethyl 3-amino-4-(methylamino)benzoate
Synonyms: ethyl 3-amino-4-(methylamino)benzoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: ZPJHHBPBCFRECW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(N)=C(NC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results