CAS RN 693-72-1
CAS RN 693-72-1
IUPAC Name:
(11E)-octadec-11-enoic acid
Synonyms:
trans-vaccenic acid TVA 11(E)-vaccenic acid octadec-11(E)-enoic acid trans-11-octadecensaeure
Molecular Weight (g/mol):
282.47
Molecular Formula:
C18H34O2
InChi Key:
UWHZIFQPPBDJPM-BQYQJAHWSA-N
SMILES:
CCCCCC\C=C\CCCCCCCCCC(O)=O
1
–
1
of
1
results
1
trans-Vaccenic acid, MedChemExpress
MedChemExpress trans-Vaccenic acid is a precursor for the synthesis of saturated fatty acid in the rumen and of conjugated linoleic acid (CLA) at the tissue level.