CAS RN 878143-33-0
CAS RN 878143-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dolasetron Mesylate hydrate, MedChemExpress
MedChemExpress Dolasetron Mesylate hydrate (MDL-73147EF hydrate) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.