311.41 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
Use promo code "21665" to get your promotional price.
Quantity | 25 L |
---|---|
Packaging | Plastic drum |
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Chemical Name or Material | Acetic acid |
---|---|
Melting Point | 16.0°C to 16.5°C |
CAS | 64-19-7 |
Boiling Point | 117.0°C to 118.0°C |
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Assay Percent Range | 99.5% |
Molecular Formula | C2H4O2 |
Linear Formula | CH3COOH |
MDL Number | MFCD00036152 |
Beilstein | 02, 96 |
Fieser | 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 |
Merck Index | 15, 54 |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
Formula Weight | 60.05 |
Percent Purity | 99.5% |
Grade | Pure |
Density | 1.0480g/mL |
Quantity | 25 L |
Flash Point | 40°C |
Infrared Spectrum | Authentic |
Packaging | Plastic drum |
Physical Form | Liquid |
Refractive Index | 1.3700 to 1.3730 |
Specific Gravity | 1.048 |