Learn More
0.0 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
Brand:  Reagecon CRSR-0-500
Product Code. 11409159
  • £127.00 / 500mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Turbidity Reference Standard, Turbidity Reference Material.
  • Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
  • Certified
  • NIST traceable
  • Non-toxic, non-carcinogenic
  • Highly accurate
  • Stable
  • EPA approved
Specifications
Specifications
Non-Ratio Instrumentation
Accuracy ± 1% NTU
0.0 NTU ± 1%
500 mL