Learn More
0.01 M Phosphoric Acid and 0.01 M Monobasic Sodium Phosphate Buffered Eluent, pH 2.5, In Accordance with United States Pharmacopoeia (USP), Reagecon
Guaranteed stability throughout entire shelf life even after opening the bottle, NIST traceable
Brand: Reagecon USP811
Code : A0
Additional Details : Weight : 1.00000kg
Specifications
0.01 M | |
1 L | |
Mobile Phase Eluent |
pH 2.5 buffered eluent | |
Liquid |