missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, strips of 8

Product Code. 10394223
Click to view available options
Color:
Blue
Green
Natural
Purple
Red
Closure Type:
Domed Cap
Not Included
Product Type:
8-Strip Cap
8-Strip Tube
Quantity:
10 Packs of 12 Strips
250 Strips
Unit Size:
Pack of 120
Pack of 250
This item is not returnable. View return policy

Product Code. 10394223

Brand: Thermo Scientific™ AB0266/B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified free from human DNA, RNases, and DNases
Color Blue
Volume (Metric) 0.2 mL
Quantity 250 Strips
Shape V-Bottom
Material Polypropylene
Closure Type Domed Cap
Product Type 8-Strip Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.