missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tubes, strips of 8
Click to view available options
Color:
Blue
Green
Natural
Purple
Red
Closure Type:
Domed Cap
Not Included
Product Type:
8-Strip Cap
8-Strip Tube
Quantity:
10 Packs of 12 Strips
250 Strips
Unit Size:
Pack of 120
Pack of 250
Descripción
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
- Compatible with 0.2 mL thermal cycler blocks
- 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Especificaciones
Especificaciones
| Certifications/Compliance | Certified free from human DNA, RNases, and DNases |
| Color | Green |
| Volume (Metric) | 0.2 mL |
| Quantity | 10 Packs of 12 Strips |
| Shape | V-Bottom |
| Material | Polypropylene |
| Closure Type | Domed Cap |
| Product Type | 8-Strip Tube |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido