missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tubes, strips of 8,white
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Brand: Thermo Scientific™ AB0264W
115.46 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
- Compatible with 0.2 mL thermal cycler blocks
- 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications
Certified DNA/RNase Free | |
0.2 mL | |
250 strips | |
Polypropylene | |
Not Included | |
8-Strip Tube |
White | |
0.25 mL | |
Thermal Cyclers | |
Standard | |
250 Strip Tubes |