missing translation for 'onlineSavingsMsg'
Learn More

0.2 NTU Non-Ratio Turbidity Standard, Reagecon
BIOPHARMA

Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.

Brand:  Reagecon CRS-0.2-100

Product Code. 11516603

  • £75.85 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Turbidity Reference Standard, Turbidity Reference Material.
  • Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
  • Certified
  • NIST traceable
  • Non-toxic, non-carcinogenic
  • Highly accurate
  • Stable
  • EPA approved
Specifications

Specifications

Non Ratio Instruments
Liquid format in an HDPE bottle
0.2 NTU ± 1%
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCRS0224B1 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog NumberCRS-0.2-100
Lot NumberCRS0223E1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberCRS-0.2-100
Lot NumberCRS0222G1 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog NumberCRS-0.2-100
Lot NumberCRS0220M1 Certificate TypeCertificate of Analysis Date26/04/2022 Catalog NumberCRS-0.2-100
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
0.2 NTU Non-Ratio Turbidity Standard, Reagecon > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.