missing translation for 'onlineSavingsMsg'
Learn More
Learn More
0.5 mg/L C as Sucrose Total Organic Carbon (TOC) Standard for Sievers 900/M9 Analyzers, Reagecon
Extremely pure and specific product made from ultra-pure raw materials using a proprietary process.
Brand: Reagecon RTOCS01
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Total Organic/Inorganic Carbon (TOC/TIC) Reference Standard, Total Organic/Inorganic Carbon (TOC/TIC) Reference Material.
- Extremely high specification and purity.
- Produced from ultra pure raw materials, including water that is prepared by a special proprietary process.
- Presented in vials that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.
- Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
- Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
- The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
- The concentration of these standards is verified using a high performance calibrated Total Organic Carbon Analyzer (TOC Instrument) using Reagecon’s ISO 17025 accredited test method (INAB Ref: 264T).
- The calibration of the TOC instrument is completed using a high-purity ISO Guide 34 accredited TOC standard similar in TOC concentration value to this product.
Specifications
Accuracy: ± 2%, Matrix: water | |
TOC Glass Vial with Septa & Cap | |
TOC Analysis |
0.5 mg/L | |
35 mL |