missing translation for 'onlineSavingsMsg'
Learn More

0.5 mg/L C from USP Sucrose Rs Total Organic Carbon (TOC) Standard for Sievers 900/M9 Analyzers, Reagecon
BIOPHARMA

Extremely pure and specific product made from ultra-pure raw materials using a proprietary process.

Brand:  Reagecon RTOCRS

Product Code. 14822171

  • £82.55 / 35mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Total Organic/Inorganic Carbon (TOC/TIC) Reference Standard, Total Organic/Inorganic Carbon (TOC/TIC) Reference Material.
  • Extremely high specification and purity.
  • Produced from ultra pure raw materials, including water that is prepared by a special proprietary process.
  • Presented in vials that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.
  • Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
  • Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
  • The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
  • The concentration of these standards is verified using a high performance calibrated Total Organic Carbon Analyzer (TOC Instrument) using Reagecon’s ISO 17025 accredited test method (INAB Ref: 264T).
  • The calibration of the TOC instrument is completed using a high-purity ISO Guide 34 accredited TOC standard similar in TOC concentration value to this product.
Specifications

Specifications

Accuracy: ± 2%, Matrix: water
35 mL
TOC Analysis
TOC Glass Vial with Septa & Cap
0.5 mg/L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberTOCRS25B1 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberRTOCRS
Lot NumberTOCRS24L3 Certificate TypeCertificate of Analysis Date10/12/2024 Catalog NumberRTOCRS
Lot NumberTOCRS23C2 Certificate TypeCertificate of Analysis Date30/03/2023 Catalog NumberRTOCRS
Lot NumberTOCRS22L2 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog NumberRTOCRS
Lot NumberTOCRS22K2 Certificate TypeCertificate of Analysis Date14/11/2022 Catalog NumberRTOCRS
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
0.5 mg/L C from USP Sucrose Rs Total Organic Carbon (TOC) Standard for Sievers 900/M9 Analyzers, Reagecon > 35 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.