missing translation for 'onlineSavingsMsg'
Learn More

Labcon 0.5 mL SuperSlik™ Low Retention Microcentrifuge Tubes with Attached Caps
GREENER_CHOICE

Product Code. 16624012
Click to view available options
Capacity (Metric):
0.5 mL
Unit Size:
Pack of 500

Product Code. 16624012

Brand: Labcon 30255700009

Only null left
Add to basket
Add to basket

With flat snap caps, side graduations, and frosted writing surface

  • Microcentrifuge tube
  • With flat snap caps, side graduations, and frosted writing surface
  • Ultra low fluid retention
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • High centrifuging to 35,000 x G
  • Freezable to -80°C
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Autoclavable Autoclavable
Capacity (Metric) 0.5 mL
Color Natural
Material Polypropylene Resin
For Use With (Application) Centrifugation
Packaging 500 / resealable bag 10 bags / case
Volume (Metric) 0.5 mL
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Sterility Non-sterile
Product Line SuperSlik™ Low Retention microcentrifuge tubes
Graduated Yes
Max. RCF 20,000 RCF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.