missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amber Screw Cap Micro Tubes
GREENER_CHOICE

Product Code. 16205570 Shop All MBP Molecular BioProducts Products
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Bottom Shape:
Conical
Non-skirted
Skirted
Closure Type:
Attached Cap
Not Included
Screw Cap with O-ring (Attached)
Screw Cap with O-ring (Unattached)
Packaging:
50 tubes and attached caps/bag
50 tubes/bag, 10 bags/unit, 10 units/case; 500 caps/bag/unit
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Unit Size:
Pack of 500
Pack of 5000
This item is not returnable. View return policy

Product Code. 16205570

Brand: Thermo Scientific™ 3470ANKS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.

Thermo Scientific™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes. Options are available with or without a self-standing skirted bottom, non-knurl and knurled tubes for single hand operation, in 0.5mL, 1.5mL and 2.0mL sizes, and the option of multicolored caps, with or without tethered straps. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.

Note: For screw caps, see Cat. No. 3471AS.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 2 mL
Color Amber
Material Polypropylene (Virgin)
Includes NonKnurl
Packaging 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric) Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Clean Claims Sterile Electron Beam, SAL 10-6 Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, Bioburden, ATP & PCR Inhibitor
UV Resistance (UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm
Binding Property Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Max. Torque Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Sterility Sterile
Closure Type Not Included
Fit for Purpose Sterile and PCR Ready
Bottom Shape Non-skirted
Max. RCF 20,000 x g
Product Type Screw-cap Microtube
Show More Show Less

† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.

Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.

Not intended to submerge and store in liquid phase LN2.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.