Learn More
Thermo Scientific™ Amber Screw Cap Micro Tubes
Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.
Brand: Thermo Scientific™ 3466AS
359.65 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 6.33000kg
Description
Thermo Scientific™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes. Options are available with or without a self-standing skirted bottom, non-knurl and knurled tubes for single hand operation, in 0.5mL, 1.5mL and 2.0mL sizes, and the option of multicolored caps, with or without tethered straps. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Note: For screw caps, see Cat. No. 3471AS.Specifications
20,000 x g | |
No | |
Amber | |
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag. | |
Sterile Electron Beam, SAL 10-6 Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, Bioburden, ATP & PCR Inhibitor | |
Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition | |
Sterile | |
Sterile & PCR Ready |
1.5ml Screw Cap Tube,Knurl, NonSkirted,Amber, E-Beam Sterile, tube only no cap | |
1.5 mL | |
Polypropylene (Virgin) | |
Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage | |
(UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm | |
Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm | |
No |
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.