Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
£47.64 - £3279.32
Chemical Identifiers
CAS | 920-66-1 |
---|---|
MDL Number | MFCD00011651 |
InChI Key | BYEAHWXPCBROCE-UHFFFAOYSA-N |
Synonym | 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcoholShow More |
PubChem CID | 13529 |
ChEBI | CHEBI:63104 |
IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-ol |
SMILES | C(C(F)(F)F)(C(F)(F)F)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10501971
|
Thermo Scientific Acros
147540050 |
5 g | Glass bottle |
£47.64
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205200
|
Thermo Scientific Acros
147540250 |
25 g | Glass Bottle |
£95.78
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10706191
|
Thermo Scientific Acros
147541000 |
100 g | Glass Bottle |
£245.22
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499623
|
Thermo Scientific Acros
147545000 |
500 g | Glass bottle |
£903.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15284634
|
Thermo Scientific Acros
147540025 |
2.5 kg | Glass bottle |
£3279.32
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
920-66-1 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
13529 | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcoholShow More | |
CHEBI:63104 | |
C(C(F)(F)F)(C(F)(F)F)O |
Specifications
920-66-1 | |
100.0 | |
Colorless | |
59.0°C | |
Authentic | |
C3H2F6O | |
1.2740 to 1.2770 | |
1.596 | |
Solubility in water: 1000g/L (25°C). | |
C(C(F)(F)F)(C(F)(F)F)O | |
168.04 | |
CHEBI:63104 | |
168.04 | |
99.5+% | |
Liquid |
99.5 | |
-4.0°C | |
1.5960g/mL | |
>100°C | |
99.5% min. (GC) | |
(CF3)2CHOH | |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
13529 | |
1.65 mPa.s (20°C) | |
102mmHg at 20°C | |
Pure | |
1,1,1,3,3,3-Hexafluoro-2-propanol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-059-4
- 1,1,1,3,3,3-Hexafluoro-2-propanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.