missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Melting Point | 18.0°C to 21.0°C |
Density | 1.5800g/mL |
Boiling Point | 105.0°C to 108.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 25 g |
Specific Gravity | 1.58 |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 166.02 |
Percent Purity | 98% |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Hexafluoroacetone hydrate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'