missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™

Product Code. 11433801
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents.

  • 1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents, such as:
    • Polyamides, polyacrylonitriles, polyacetals, polyesters (e.g. polyglycolide), and polyketones.
  • It has also found use in biochemistry to solubilize peptides and to monomerize β-sheet protein aggregates.
  • It can be used as an acid in volatile buffers for ion pair HPLC - mass spectrometry of nucleic acids.
  • 1,1,1,3,3,3-Hexafluoro-2-propanol was used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29463-77-2
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Color Colorless
Density 1.62 g/mL
Assay Percent Range ≥99%
Quantity 100 mL
Formula Weight 168.04
Percent Purity 0.9969
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-isopropanol, For GC analysis

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'