missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2,4-hexanedione, 70%, tech., Thermo Scientific™

Product Code. 10761325
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 400-54-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00051306
InChI Key SBRYTXXHTKCMFT-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,4-hexanedione, 2,4-hexanedione, 1,1,1-trifluoro, 1-trifluoroacetyl butan-2-one, 1,1,1-trifluorohexan-2,4-dione, 2,4-dioxo-1,1,1-trifluorohexane, 1,1,1-trifluoro-2,4-hexane-dione, 1,1,1-trifluoro-hexane-2,4-dione, 1,1,1,-trifluoro-2,4-hexanedione, 2,4-hexanedione,1,1,1-trifluoro
PubChem CID 67864
IUPAC Name 1,1,1-trifluorohexane-2,4-dione
SMILES CCC(=O)CC(=O)C(F)(F)F

Specifications

CAS Min % 65.0
CAS Max % 100.0
Color Yellow
Density 28g/mL
Boiling Point 125.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 65% min. (GC)
Packaging Glass Bottle
Linear Formula CF3COCH2COCH2CH3
Refractive Index 1.3931 to 1.3951
Quantity 100 g
Formula Weight 168.12
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-2,4-hexanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1-Trifluoro-2,4-hexanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'