missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2,4-pentanedione, 98%, Thermo Scientific Chemicals

Product Code. 10206850
Click to view available options
Quantity:
10 mL
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-57-7
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00000427
InChI Key SHXHPUAKLCCLDV-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
PubChem CID 73943
IUPAC Name 1,1,1-trifluoropentane-2,4-dione
SMILES CC(=O)CC(=O)C(F)(F)F

Specifications

Color Orange to Yellow
Density 1.2700g/mL
Boiling Point 105.0°C to 107.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2COCF3
Refractive Index 1.3870 to 1.3900
Quantity 25 mL
Beilstein 01,III,3123
Specific Gravity 1.27
Solubility Information Solubility in water: slightly soluble
Formula Weight 154.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-2,4-pentanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1-Trifluoro-2,4-pentanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'