missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoro-2,4-pentanedione, 98%, Thermo Scientific Chemicals
£51.30 - £380.00
Chemical Identifiers
CAS | 367-57-7 |
---|---|
Molecular Formula | C5H5F3O2 |
Molecular Weight (g/mol) | 154.09 |
MDL Number | MFCD00000427 |
InChI Key | SHXHPUAKLCCLDV-UHFFFAOYSA-N |
Synonym | 1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone |
PubChem CID | 73943 |
IUPAC Name | 1,1,1-trifluoropentane-2,4-dione |
SMILES | CC(=O)CC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10286840
|
Thermo Scientific Acros
216710100 |
10 mL |
£51.30
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10206850
|
Thermo Scientific Acros
216710250 |
25 mL |
£104.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11994871
|
Thermo Scientific Acros
216711000 |
100 mL |
£380.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
367-57-7 | |
154.09 | |
SHXHPUAKLCCLDV-UHFFFAOYSA-N | |
73943 | |
CC(=O)CC(=O)C(F)(F)F |
C5H5F3O2 | |
MFCD00000427 | |
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone | |
1,1,1-trifluoropentane-2,4-dione |
Specifications
367-57-7 | |
1.2700g/mL | |
26°C | |
97.5% min. (GC) | |
C5H5F3O2 | |
1.3870 to 1.3900 | |
01,III,3123 | |
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone | |
SHXHPUAKLCCLDV-UHFFFAOYSA-N | |
1,1,1-trifluoropentane-2,4-dione | |
73943 | |
98% | |
1,1,1-Trifluoro-2,4-pentanedione |
Orange to Yellow | |
105.0°C to 107.0°C | |
Authentic | |
Glass bottle | |
CH3COCH2COCF3 | |
MFCD00000427 | |
1.27 | |
Solubility in water: slightly soluble | |
CC(=O)CC(=O)C(F)(F)F | |
154.09 | |
154.09 | |
Liquid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-698-5
Recommended Storage : Product may darken in storage
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,1,1-Trifluoro-2,4-pentanedione
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only