missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
20 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 1.6100g/mL |
Deuterium (D) | 98.5% min. |
Packaging | Glass Bottle |
Refractive Index | 1.4924 |
Quantity | 5 mL |
Boiling Point | 146.0°C |
Linear Formula | (Cl2CD)2 |
Beilstein | 01, III, 164 |
Show More |
Safety and Handling
Product Identifier
- 1,1,2,2-Tetrachloroethane-d2
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H310-Fatal in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'