missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2-Trichloroethane, 98%, Thermo Scientific Chemicals
£67.70 - £411.00
Chemical Identifiers
CAS | 79-00-5 |
---|---|
Molecular Formula | C2H3Cl3 |
Molecular Weight (g/mol) | 133.40 |
MDL Number | MFCD00000852 |
InChI Key | UBOXGVDOUJQMTN-UHFFFAOYSA-N |
Synonym | ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t |
PubChem CID | 6574 |
ChEBI | CHEBI:36018 |
IUPAC Name | 1,1,2-trichloroethane |
SMILES | ClCC(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447720
|
Thermo Scientific Acros
139432500 |
250 mL | Glass bottle |
£67.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10427720
|
Thermo Scientific Acros
139430010 |
1 L | Glass bottle |
£192.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974471
|
Thermo Scientific Acros
139430025 |
2.5 L | Glass bottle |
£411.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-00-5 | |
133.40 | |
UBOXGVDOUJQMTN-UHFFFAOYSA-N | |
6574 | |
1,1,2-trichloroethane |
C2H3Cl3 | |
MFCD00000852 | |
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t | |
CHEBI:36018 | |
ClCC(Cl)Cl |
Specifications
79-00-5 | |
Colorless | |
110.0°C to 115.0°C | |
C2H3Cl3 | |
1.4700 to 1.4720 | |
01,85 | |
1.43 | |
Solubility in water: 4g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,miscible with alcohols,ethers,esters and ketones | |
ClCC(Cl)Cl | |
133.40 | |
CHEBI:36018 | |
133.4 | |
Liquid |
-37.0°C | |
1.4300g/mL | |
97.5% min. (GC) | |
ClCH2CHCl2 | |
MFCD00000852 | |
15,9802 | |
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t | |
UBOXGVDOUJQMTN-UHFFFAOYSA-N | |
1,1,2-trichloroethane | |
6574 | |
1.69 mPa.s (25°C) | |
98% | |
1, 1, 2-Trichloroethane |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-166-9
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
Hazard Statement
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only