missing translation for 'onlineSavingsMsg'
Learn More

1,1':3',1″-Terphenyl-5'-boronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15483737
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

Solubility
Insoluble in water.

Notes
Refrigerated (2-4°C). Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128388-54-5
Molecular Formula C18H15BO2
Molecular Weight (g/mol) 274.126
MDL Number MFCD09953491
InChI Key MRBZYVMZUBUDAX-UHFFFAOYSA-N
Synonym 3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x
PubChem CID 14739363
IUPAC Name (3,5-diphenylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 292°C to 296°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 274.13
Percent Purity 95%
Chemical Name or Material 1,1':3',1''-Terphenyl-5'-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'