missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetramethyldisiloxane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
Boiling Point | 70.0°C to 71.0°C |
Melting Point | -78.0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 mL |
Assay Percent Range | 97% |
Linear Formula | [(CH3)2SiH]2O |
Flash Point | −20°C |
Solubility Information | Solubility in water: decomposes. Other solubilities: decomposes with alcohol |
Formula Weight | 134.33 |
Show More |
Safety and Handling
Product Identifier
- 1,1,3,3-Tetramethyldisiloxane
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'