Learn More
1,1,3,3-Tetramethylguanidine, 99%, Thermo Scientific Chemicals
£30.50 - £329.00
Chemical Identifiers
CAS | 80-70-6 |
---|---|
Molecular Weight (g/mol) | 115.18 |
InChI Key | KYVBNYUBXIEUFW-UHFFFAOYSA-N |
Synonym | tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine |
PubChem CID | 66460 |
SMILES | CN(C)C(=N)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10513391
|
Thermo Scientific Acros
221291000 |
100 g | Glass bottle |
£30.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276990
|
Thermo Scientific Acros
221295000 |
500 g | Glass bottle |
£116.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30117284
|
Thermo Scientific Acros
221290020 |
£329.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11995591
|
Thermo Scientific Acros
221290025 |
2.5 kg | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80-70-6 | |
KYVBNYUBXIEUFW-UHFFFAOYSA-N | |
66460 |
115.18 | |
tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine | |
CN(C)C(=N)N(C)C |
Specifications
80-70-6 | |
Colorless to Yellow | |
160.0°C to 162.0°C | |
Authentic | |
C5H13N3 | |
1.4670 to 1.4690 | |
01,1145; 04,489; 12,477 | |
0.91 | |
Solubility in water: miscible. Other solubilities: miscible | |
CN(C)C(=N)N(C)C | |
115.18 | |
0.002 Pas (20°C) | |
99% | |
1,1,3,3-Tetramethylguanidine |
-30.0°C | |
0.9100g/mL | |
50°C | |
98.5% min. (GC) | |
(CH3)2NC(=NH)N(CH3)2 | |
04,I,335 | |
15,9375 | |
tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine | |
KYVBNYUBXIEUFW-UHFFFAOYSA-N | |
1,1,3,3-tetramethylguanidine | |
66460 | |
115.18 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-302-7
RUO – Research Use Only