missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Thermo Scientific™

Product Code. 10511692
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 40288-65-1
Molecular Formula C9H7BrO3
Molecular Weight (g/mol) 243.056
InChI Key QBXCVQVFPVXAGS-UHFFFAOYSA-N
PubChem CID 243777
IUPAC Name 1-(1,3-benzodioxol-5-yl)-2-bromoethanone
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 1-1,3-benzodioxol-5-yl-2-bromoethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 5-bromoacetyl-1,3-benzodioxolane, ethanone,1-1,3-benzodioxol-5-yl-2-bromo, 1-benzo d 1,3 dioxol-5-yl-2-bromoethanone, 1-benzo 1,3 dioxol-5-yl-2-bromo-ethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethanone, ethanone, 1-1,3-benzodioxol-5-yl-2-bromo, 1-2h-benzo 3,4-d 1,3-dioxolen-5-yl-2-bromoethan-1-one
Formula Weight 243.06
Percent Purity 97%
Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'