missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 1-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 1-1,3-benzodioxol-5-yl-2-bromoethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 5-bromoacetyl-1,3-benzodioxolane, ethanone,1-1,3-benzodioxol-5-yl-2-bromo, 1-benzo d 1,3 dioxol-5-yl-2-bromoethanone, 1-benzo 1,3 dioxol-5-yl-2-bromo-ethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethanone, ethanone, 1-1,3-benzodioxol-5-yl-2-bromo, 1-2h-benzo 3,4-d 1,3-dioxolen-5-yl-2-bromoethan-1-one |
Formula Weight | 243.06 |
Percent Purity | 97% |
Chemical Name or Material | 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'