missing translation for 'onlineSavingsMsg'
Learn More
1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™ Product Code.: 10147082

1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™

Product Code. 10147082
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10147082

Brand: Thermo Scientific Maybridge KM10326DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-39-7
Molecular Formula C11H7F3O4
Molecular Weight (g/mol) 260.168
MDL Number MFCD01570563
InChI Key FDLJUCOQPAGQNY-UHFFFAOYSA-N
Synonym 1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dioneShow MoreShow Less
PubChem CID 2775368
IUPAC Name 1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 260.17
Percent Purity 97%
Chemical Name or Material 1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.