missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone, 90+%, Thermo Scientific™

Product Code. 10555144
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 54223-20-0
Molecular Formula C9H6BrNOS
Molecular Weight (g/mol) 256.12
MDL Number MFCD03659698
InChI Key AYWGYNKWZWBMSV-UHFFFAOYSA-N
Synonym 1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
PubChem CID 2776256
SMILES BrCC(=O)C1=NC2=CC=CC=C2S1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 1-(1,3-benzothiazol-2-yl)-2-bromoethan-1-one
Formula Weight 256.12
Percent Purity 90%
Chemical Name or Material 1-(1, 3-Benzothiazol-2-yl)-2-bromo-1-ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'