Learn More
1,1,3-Triethoxypropane, 98+%, Thermo Scientific™
£16.07 - £64.29
Chemical Identifiers
CAS | 7789-92-6 |
---|---|
Molecular Formula | C9H20O3 |
Molecular Weight (g/mol) | 176.26 |
MDL Number | MFCD00009241 |
InChI Key | LGICWIVABSMSDK-UHFFFAOYSA-N |
Synonym | 3-ethoxypropionaldehyde diethyl acetal, propane, 1,1,3-triethoxy, 1,3,3-triethoxypropane, 3-ethoxypropanal diethyl acetal, propane, 1,3,3-triethoxy, unii-1qw8j6vxam, 3-ethoxypropionaldehydediethylacetal, 1qw8j6vxam, beta-ethoxypropionaldehyde diethyl acetal, .beta.-ethoxypropionaldehyde diethyl acetal |
PubChem CID | 24624 |
IUPAC Name | 1,1,3-triethoxypropane |
SMILES | CCOCCC(OCC)OCC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7789-92-6 | |
176.26 | |
LGICWIVABSMSDK-UHFFFAOYSA-N | |
24624 | |
CCOCCC(OCC)OCC |
C9H20O3 | |
MFCD00009241 | |
3-ethoxypropionaldehyde diethyl acetal, propane, 1,1,3-triethoxy, 1,3,3-triethoxypropane, 3-ethoxypropanal diethyl acetal, propane, 1,3,3-triethoxy, unii-1qw8j6vxam, 3-ethoxypropionaldehydediethylacetal, 1qw8j6vxam, beta-ethoxypropionaldehyde diethyl acetal, .beta.-ethoxypropionaldehyde diethyl acetal | |
1,1,3-triethoxypropane |
Specifications
7789-92-6 | |
184°C to 186°C | |
C9H20O3 | |
MFCD00009241 | |
1098506 | |
LGICWIVABSMSDK-UHFFFAOYSA-N | |
1,1,3-triethoxypropane | |
24624 | |
≥98% |
0.898 | |
53°C (127°F) | |
1.406 | |
UN3271 | |
3-ethoxypropionaldehyde diethyl acetal, propane, 1,1,3-triethoxy, 1,3,3-triethoxypropane, 3-ethoxypropanal diethyl acetal, propane, 1,3,3-triethoxy, unii-1qw8j6vxam, 3-ethoxypropionaldehydediethylacetal, 1qw8j6vxam, beta-ethoxypropionaldehyde diethyl acetal, .beta.-ethoxypropionaldehyde diethyl acetal | |
CCOCCC(OCC)OCC | |
176.26 | |
176.26 | |
1,1,3-Triethoxypropane |
Safety and Handling
GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
missing translation for 'einecsNumber' : 232-193-4
missing translation for 'rtecsNumber' : TZ9450000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures