missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3-Trimethylcyclohexane, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,1,3-Trimethylcyclohexane | 3073-66-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 3073-66-3, 67-56-1 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.1, 99.9 |
Quantity | 1 mL |
CAS Max % | 0.1, 99.9 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Molecular Formula | C9H18 |
Solvent or Matrix | Methanol-P&T |
InChI Key | PYOLJOJPIPCRDP-UHFFFAOYSA-N |
Show More |
Safety and Handling



Signal Word
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1,1,3-Trimethylcyclohexane, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction