missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Adamantyl)ethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
Solubility
Freely soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).
Solubility
Freely soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 375°C |
Quantity | 5 g |
Merck Index | 148224 |
Solubility Information | Freely soluble in water. |
Formula Weight | 215.77 |
Percent Purity | 98% |
Chemical Name or Material | 1-(1-Adamantyl)ethylamine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'