missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 5 g |
Formula Weight | 239.07 |
Percent Purity | 95% |
Chemical Name or Material | 1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'