missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 235.0°C to 238.0°C |
Color | Yellow to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 250 mg |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 824.39 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'