missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(dicyclohexylphosphino)ferrocene, 98%, Thermo Scientific Chemicals

Product Code. 11357189
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146960-90-9
Molecular Formula C34H52FeP2
Molecular Weight (g/mol) 578.58
MDL Number MFCD09038820
IUPAC Name 1,1'-Bis(dicyclohexylphosphino)ferrocene
SMILES [Fe].C1CCC(CC1)P(C1CCCCC1)c1cccc1.C1CCC(CC1)P(C1CCCCC1)c1cccc1

Specifications

Melting Point 134°C to 136°C
Quantity 1 g
Formula Weight 578.57
Percent Purity 98%
Chemical Name or Material 1,1'-Bis(dicyclohexylphosphino)ferrocene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.