missing translation for 'onlineSavingsMsg'
Learn More
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals Product Code.: 11417686

1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals

Product Code. 11417686
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11417686

Brand: Thermo Scientific Alfa Aesar A14688.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Peptide coupling reagent1,1'-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Peptide coupling reagent1,1′-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

Solubility
Soluble in dimethylformamide.

Notes
Store in a cool place. Incompatible with water, strong acids, strong oxidizing agents, strong bases and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 116°C to 120°C
pH 7.7
Assay Percent Range 97%
Quantity 10 g
UN Number UN3263
Beilstein 6826
Sensitivity Moisture sensitive
Merck Index 14,1819
Solubility Information Soluble in dimethylformamide.
IUPAC Name di(imidazol-1-yl)methanone
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1,1'-Carbonyldiimidazole
Show More Show Less
compliance-icons
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.