missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Cyclohexanediacetic anhydride, Thermo Scientific™
£33.40 - £89.40
Specifications
Alpha Vector | CYCLOHEXANEDIACETICANHYDRIDE |
---|---|
Chemical Name or Material | 1,1-Cyclohexanediacetic anhydride |
CAS | 1010-26-0 |
Melting Point | 67°C to 72°C |
Color | Orange to White |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17780629
|
Thermo Scientific Acros
358130050 |
5 g |
£33.40
5g |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17790629
|
Thermo Scientific Acros
358130250 |
25 g |
£89.40
25g |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CYCLOHEXANEDIACETICANHYDRIDE | |
1010-26-0 | |
Orange to White | |
Glass bottle | |
00040292 | |
182.22 |
1,1-Cyclohexanediacetic anhydride | |
67°C to 72°C | |
Conforms | |
C10 H14 O3 | |
3-Oxaspiro[5.5 ]undecane-2,4-dione | |
Crystalline Powder or Crystals |