missing translation for 'onlineSavingsMsg'
Learn More

1,1-Di(methylthio)-2-nitroeth-1-ene, 97%, Thermo Scientific™

Catalog Number 10797115
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 13623-94-4
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C4H7NO2S2
Linear Formula O2NCH=C(SCH3)2
MDL Number MFCD00010443
Quantity 10 g
Synonym 1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene
InChI Key NXGHEDHQXXXTTP-UHFFFAOYSA-N
SMILES CSC(=C[N+](=O)[O-])SC
IUPAC Name 1,1-bis(methylsulfanyl)-2-nitroethene
Molecular Weight (g/mol) 165.225
PubChem CID 83623
Formula Weight 165.24
Percent Purity 97%
Chemical Name or Material 1, 1-Di(methylthio)-2-nitroeth-1-ene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'