missing translation for 'onlineSavingsMsg'
Learn More

1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil

Product Code. p-102162 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
250 mL
Unit Size:
10mL
250mL
This item is not returnable. View return policy

Product Code. 10476442

Brand: Thermo Scientific Acros 361310100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3006-86-8 | C14H28O4 | 260.374 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1, 1-Di(tert-butylperoxy)cyclohexane
Color Undesignated
Name Note 50% Solution in Mineral Oil
CAS 3006-86-8
CAS Min % 49.0
CAS Max % 51.0
Molecular Formula C14H28O4
MDL Number MFCD00084800
Beilstein 07, III, 30
Density 0.9500g/mL
Synonym vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene
InChI Key HSLFISVKRDQEBY-UHFFFAOYSA-N
SMILES CC(C)(C)OOC1(CCCCC1)OOC(C)(C)C
IUPAC Name 1,1-dichloroethene
Molecular Weight (g/mol) 260.37
PubChem CID 6366
ChEBI CHEBI:34031
Formula Weight 260.37
Percent Purity 49 to 51%
Packaging Plastic bottle
Quantity 10 mL
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.